Product Details

SNP ID
rs140110188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:681478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTTGCGGCAACAACAACGTCAC[C/T]TACATCTCCTCGTGCCACATGCGCC
Phenotype
MIM: 605343
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FSTL3 PubMed Links

Gene Details

Gene
FSTL3
Gene Name
follistatin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005860.2 686 Silent Mutation ACC,ACT T217T NP_005851.1
Gene
PRSS57
Gene Name
protease, serine 57
There are no transcripts associated with this gene.

View Full Product Details