Product Details

SNP ID
rs140418323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51032012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCAGCCGCAGCAGCCGGAGGTC[G/T]TGCTCGTGGCTCGTCGAGGCTCCCA
Phenotype
MIM: 604434 MIM: 605539
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KLK11 PubMed Links

Gene Details

Gene
KLK11
Gene Name
kallikrein related peptidase 11
There are no transcripts associated with this gene.

Gene
KLK12
Gene Name
kallikrein related peptidase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019598.2 438 Silent Mutation NP_062544.1
NM_145894.1 438 Silent Mutation NP_665901.1
NM_145895.1 438 Intron NP_665902.1
XM_005258950.3 438 Intron XP_005259007.1
XM_005258951.3 438 Intron XP_005259008.1

View Full Product Details