Product Details

SNP ID
rs140517797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38900179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGGGGCTGTCGTGGGCTCCCCT[C/T]GTCTTCGGCTACGTCACTGAGGATG
Phenotype
MIM: 604495 MIM: 604480
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCER2 PubMed Links

Gene Details

Gene
CCER2
Gene Name
coiled-coil glutamate rich protein 2
There are no transcripts associated with this gene.

Gene
NFKBIB
Gene Name
NFKB inhibitor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243116.1 242 Intron NP_001230045.1
NM_002503.4 242 Silent Mutation CTC,CTT L49L NP_002494.2
XM_006723226.2 242 Missense Mutation TCG,TTG S36L XP_006723289.2
XM_006723227.3 242 Intron XP_006723290.3
Gene
SIRT2
Gene Name
sirtuin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193286.1 242 Intron NP_001180215.1
NM_012237.3 242 Intron NP_036369.2
NM_030593.2 242 Intron NP_085096.1
XM_006723111.1 242 Intron XP_006723174.1
XM_011526654.1 242 Intron XP_011524956.1
XM_011526655.1 242 Intron XP_011524957.1
XM_017026500.1 242 Intron XP_016881989.1

View Full Product Details