Product Details

SNP ID
rs140748603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19527293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCTTTCACAGGCGCCTACAAAT[C/T]GAGGACTTCGAGGCTCGCATCGCGC
Phenotype
MIM: 609435
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFA13 PubMed Links

Gene Details

Gene
NDUFA13
Gene Name
NADH:ubiquinone oxidoreductase subunit A13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015965.6 215 Silent Mutation ATC,ATT I62I NP_057049.5
Gene
YJEFN3
Gene Name
YjeF N-terminal domain containing 3
There are no transcripts associated with this gene.

View Full Product Details