Product Details

SNP ID
rs140971020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39415204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTCAAGGCTGGAGCGTGTGGCC[C/T]GGGAGATCGTGGAGACAGAACGGGC
Phenotype
MIM: 611893 MIM: 190700
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4530 PubMed Links

Gene Details

Gene
MIR4530
Gene Name
microRNA 4530
There are no transcripts associated with this gene.

Gene
PLEKHG2
Gene Name
pleckstrin homology and RhoGEF domain containing G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022835.2 1178 Missense Mutation CGG,TGG R108W NP_073746.2
XM_005259163.2 1178 Missense Mutation CGG,TGG R108W XP_005259220.1
XM_006723334.2 1178 Missense Mutation CGG,TGG R49W XP_006723397.1
XM_011527232.2 1178 Missense Mutation CGG,TGG R109W XP_011525534.1
XM_017027150.1 1178 Missense Mutation CGG,TGG R109W XP_016882639.1
XM_017027151.1 1178 Missense Mutation CGG,TGG R109W XP_016882640.1
Gene
ZFP36
Gene Name
ZFP36 ring finger protein
There are no transcripts associated with this gene.

View Full Product Details