Product Details

SNP ID
rs141206410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44957824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACCCACAGCTTTGCCATTCCTTC[A/C]TTCAGCGAATGCGAATTGAGTCTAC
Phenotype
MIM: 608083 MIM: 604783
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APOC2 PubMed Links

Gene Details

Gene
APOC2
Gene Name
apolipoprotein C2
There are no transcripts associated with this gene.

Gene
APOC4-APOC2
Gene Name
APOC4-APOC2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
CLPTM1
Gene Name
CLPTM1, transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282175.1 Intron NP_001269104.1
NM_001282176.1 Intron NP_001269105.1
NM_001294.3 Intron NP_001285.1

View Full Product Details