Product Details
- SNP ID
-
rs141234591
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:48713916 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTCCAGAGTCAGAGCCGCTGAGC[C/T]CGGCCCTGAGGAAGGGGTCAGGGCT
- Phenotype
-
MIM: 182100
MIM: 610349
MIM: 609623
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FUT2
PubMed Links
Gene Details
- Gene
- FUT2
- Gene Name
- fucosyltransferase 2
There are no transcripts associated with this gene.
- Gene
- MAMSTR
- Gene Name
- MEF2 activating motif and SAP domain containing transcriptional regulator
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130915.1 |
1683 |
Missense Mutation |
AGC,GGC |
S285G |
NP_001124387.1 |
NM_001297753.1 |
1683 |
Missense Mutation |
AGC,GGC |
S117G |
NP_001284682.1 |
NM_182574.2 |
1683 |
Missense Mutation |
AGC,GGC |
S182G |
NP_872380.1 |
XM_011526807.2 |
1683 |
Missense Mutation |
AGC,GGC |
S182G |
XP_011525109.1 |
XM_011526808.2 |
1683 |
Missense Mutation |
AGC,GGC |
S348G |
XP_011525110.1 |
XM_011526809.2 |
1683 |
Missense Mutation |
AGC,GGC |
S117G |
XP_011525111.1 |
XM_017026640.1 |
1683 |
Missense Mutation |
AGC,GGC |
S366G |
XP_016882129.1 |
XM_017026641.1 |
1683 |
Missense Mutation |
AGC,GGC |
S353G |
XP_016882130.1 |
XM_017026642.1 |
1683 |
Missense Mutation |
AGC,GGC |
S182G |
XP_016882131.1 |
XM_017026643.1 |
1683 |
Missense Mutation |
AGC,GGC |
S313G |
XP_016882132.1 |
XM_017026644.1 |
1683 |
Missense Mutation |
AGC,GGC |
S303G |
XP_016882133.1 |
XM_017026645.1 |
1683 |
Missense Mutation |
AGC,GGC |
S117G |
XP_016882134.1 |
- Gene
- RASIP1
- Gene Name
- Ras interacting protein 1
There are no transcripts associated with this gene.
View Full Product Details