Product Details

SNP ID
rs141241402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57135383 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGTGTTTCACAAGATCTGACTT[G/T]TAAATGAAAGCCTTTCCACAGTCCG
Phenotype
MIM: 609546
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
USP29 PubMed Links

Gene Details

Gene
USP29
Gene Name
ubiquitin specific peptidase 29
There are no transcripts associated with this gene.

Gene
ZIM3
Gene Name
zinc finger imprinted 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052882.1 1340 Silent Mutation NP_443114.1

View Full Product Details