Product Details

SNP ID
rs141344265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55378138 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAAAGAGTCCAGGGATGTGGAG[A/G]GAGGCACCGAGGGGGAAGGGACGGA
Phenotype
MIM: 147681 MIM: 603638
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL11 PubMed Links

Gene Details

Gene
IL11
Gene Name
interleukin 11
There are no transcripts associated with this gene.

Gene
RPL28
Gene Name
ribosomal protein L28
There are no transcripts associated with this gene.

Gene
TMEM190
Gene Name
transmembrane protein 190
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139172.1 487 Missense Mutation AGA,GGA R157G NP_631911.1
XM_017026331.1 487 Missense Mutation AGA,GGA R115G XP_016881820.1
Gene
TMEM238
Gene Name
transmembrane protein 238
There are no transcripts associated with this gene.

View Full Product Details