Product Details

SNP ID
rs141496051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56441389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATAAGAGATGTGCGCTGACTAA[A/G]GGCCTTACCACATGTTTTGCATGTA
Phenotype
MIM: 611024
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF667 PubMed Links

Gene Details

Gene
ZNF667
Gene Name
zinc finger protein 667
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321355.1 1795 Missense Mutation CTT,TTT L479F NP_001308284.1
NM_001321356.1 1795 Missense Mutation CTT,TTT L536F NP_001308285.1
NM_022103.3 1795 Missense Mutation CTT,TTT L536F NP_071386.3
XM_011527208.1 1795 Missense Mutation CTT,TTT L536F XP_011525510.1
XM_011527209.1 1795 Missense Mutation CTT,TTT L536F XP_011525511.1

View Full Product Details