Product Details

SNP ID
rs141530220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44287737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCACATTTGTAGGGTTTCTCTCC[A/G]GTGTGGACTCTCTGATGATTGTGAA
Phenotype
MIM: 604749
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF235 PubMed Links

Gene Details

Gene
ZNF235
Gene Name
zinc finger protein 235
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004234.4 5857 Silent Mutation ACC,ACT T566T NP_004225.3
XM_005259412.4 5857 Silent Mutation ACC,ACT T562T XP_005259469.1

View Full Product Details