Product Details

SNP ID
rs141607224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54356361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATTCCCAGGGGCCTCACCTCTG[C/T]TGTGGCTTCTGCTCCTCGTCCTTGC
Phenotype
MIM: 602992
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LAIR1 PubMed Links

Gene Details

Gene
LAIR1
Gene Name
leukocyte associated immunoglobulin like receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289023.2 931 Silent Mutation CAA,CAG Q189Q NP_001275952.2
NM_001289025.2 931 Silent Mutation CAA,CAG Q206Q NP_001275954.2
NM_001289026.2 931 Silent Mutation CAA,CAG Q200Q NP_001275955.2
NM_001289027.2 931 Silent Mutation CAA,CAG Q189Q NP_001275956.2
NM_002287.5 931 Silent Mutation CAA,CAG Q207Q NP_002278.2
NM_021706.4 931 Silent Mutation CAA,CAG Q190Q NP_068352.2
XM_017026803.1 931 Intron XP_016882292.1

View Full Product Details