Product Details

SNP ID
rs141708238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18784290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGTCCTTCCACAGCAGCCGCACC[A/T]GGGACTCTGTGTCTCCTGTATGCCA
Phenotype
MIM: 600310 MIM: 607536
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
COMP PubMed Links

Gene Details

Gene
COMP
Gene Name
cartilage oligomeric matrix protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000095.2 2024 Missense Mutation CAG,CTG Q663L NP_000086.2
Gene
CRTC1
Gene Name
CREB regulated transcription coactivator 1
There are no transcripts associated with this gene.

View Full Product Details