Product Details

SNP ID
rs141910004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39415366 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACCTGGGCCCTCTGCTGGACGGC[A/G]GGGTCCTGGGGCTGAGCGTGGAGCA
Phenotype
MIM: 611893 MIM: 190700
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4530 PubMed Links

Gene Details

Gene
MIR4530
Gene Name
microRNA 4530
There are no transcripts associated with this gene.

Gene
PLEKHG2
Gene Name
pleckstrin homology and RhoGEF domain containing G2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022835.2 1262 Missense Mutation AGG,GGG R136G NP_073746.2
XM_005259163.2 1262 Missense Mutation AGG,GGG R136G XP_005259220.1
XM_006723334.2 1262 Missense Mutation AGG,GGG R77G XP_006723397.1
XM_011527232.2 1262 Missense Mutation AGG,GGG R137G XP_011525534.1
XM_017027150.1 1262 Missense Mutation AGG,GGG R137G XP_016882639.1
XM_017027151.1 1262 Missense Mutation AGG,GGG R137G XP_016882640.1
Gene
ZFP36
Gene Name
ZFP36 ring finger protein
There are no transcripts associated with this gene.

View Full Product Details