Product Details

SNP ID
rs142179752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11576341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGCAGTGGGTAGGCCCGTGCT[C/G]GGCTATGGACCACACGGGGTAGTGG
Phenotype
MIM: 171640 MIM: 612248
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACP5 PubMed Links

Gene Details

Gene
ACP5
Gene Name
acid phosphatase 5, tartrate resistant
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111034.2 1040 Missense Mutation CAG,GAG Q213E NP_001104504.1
NM_001111035.2 1040 Missense Mutation CAG,GAG Q213E NP_001104505.1
NM_001111036.2 1040 Missense Mutation CAG,GAG Q213E NP_001104506.1
NM_001322023.1 1040 Missense Mutation CAG,GAG Q213E NP_001308952.1
NM_001611.4 1040 Missense Mutation CAG,GAG Q213E NP_001602.1
XM_005259938.1 1040 Missense Mutation CAG,GAG Q213E XP_005259995.1
XM_011528069.2 1040 Missense Mutation CAG,GAG Q213E XP_011526371.1
Gene
ZNF627
Gene Name
zinc finger protein 627
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290083.1 1040 Intron NP_001277012.1
NM_001290084.1 1040 Intron NP_001277013.1
NM_001290085.1 1040 Intron NP_001277014.1
NM_145295.3 1040 Intron NP_660338.1
XM_011527780.2 1040 Intron XP_011526082.1

View Full Product Details