Product Details

SNP ID
rs142423982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1910034 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGCTGTTCTGGGAGGGGGCCCC[A/G]TGTCCCCCACACTTGGCATGCTTGC
Phenotype
MIM: 615302 MIM: 613764
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAT3 PubMed Links

Gene Details

Gene
ADAT3
Gene Name
adenosine deaminase, tRNA specific 3
There are no transcripts associated with this gene.

Gene
SCAMP4
Gene Name
secretory carrier membrane protein 4
There are no transcripts associated with this gene.

View Full Product Details