Product Details

SNP ID
rs142460503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35888577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGTCCTAAGAGGACAGGCCTGG[C/T]GGCGCCACACGGCTCCTCTTCAACA
Phenotype
MIM: 104775
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APLP1 PubMed Links

Gene Details

Gene
APLP1
Gene Name
amyloid beta precursor like protein 1
There are no transcripts associated with this gene.

Gene
NFKBID
Gene Name
NFKB inhibitor delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321831.1 1242 Silent Mutation NP_001308760.1
NM_139239.2 1242 Silent Mutation NP_640332.1
XM_005259340.3 1242 Silent Mutation XP_005259397.1
XM_011527419.2 1242 Silent Mutation XP_011525721.1
XM_011527420.2 1242 Silent Mutation XP_011525722.1
XM_011527421.2 1242 Silent Mutation XP_011525723.1
XM_011527424.2 1242 Silent Mutation XP_011525726.1
XM_017027389.1 1242 Intron XP_016882878.1
XM_017027390.1 1242 Silent Mutation XP_016882879.1
XM_017027391.1 1242 Silent Mutation XP_016882880.1
XM_017027392.1 1242 Silent Mutation XP_016882881.1
XM_017027393.1 1242 Silent Mutation XP_016882882.1
XM_017027394.1 1242 Silent Mutation XP_016882883.1
XM_017027395.1 1242 Silent Mutation XP_016882884.1
XM_017027396.1 1242 Intron XP_016882885.1
XM_017027397.1 1242 Silent Mutation XP_016882886.1

View Full Product Details