Product Details

SNP ID
rs142615352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56383968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGCTGATGGAAGGCTTTTCTAC[A/G]TTTATTATATTCACATGTGTTTCTC
Phenotype
MIM: 615600
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF542P PubMed Links

Gene Details

Gene
ZNF542P
Gene Name
zinc finger protein 542, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF582
Gene Name
zinc finger protein 582
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320371.1 1689 Silent Mutation AAC,AAT N514N NP_001307300.1
NM_144690.2 1689 Silent Mutation AAC,AAT N483N NP_653291.1
Gene
ZNF582-AS1
Gene Name
ZNF582 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details