Product Details

SNP ID
rs142693254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4839442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCACCTGGTCCTTCACATTGGTG[A/G]GGAGGCCCTGAATGCTGGACCCCAG
Phenotype
MIM: 602702 MIM: 607601
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLIN3 PubMed Links

Gene Details

Gene
PLIN3
Gene Name
perilipin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164189.1 1232 Missense Mutation CCC,CTC P351L NP_001157661.1
NM_001164194.1 1232 Missense Mutation CCC,CTC P340L NP_001157666.1
NM_005817.4 1232 Missense Mutation CCC,CTC P352L NP_005808.3
Gene
TICAM1
Gene Name
toll like receptor adaptor molecule 1
There are no transcripts associated with this gene.

View Full Product Details