Product Details

SNP ID
rs142976406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55402884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGGAAACCCCAATTCCCCACC[C/T]TCTCTGCCATGTGCCCCAGGGACTC
Phenotype
MIM: 610309
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE2S PubMed Links

Gene Details

Gene
UBE2S
Gene Name
ubiquitin conjugating enzyme E2 S
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014501.2 Intron NP_055316.2
XM_011526752.1 Intron XP_011525054.1

View Full Product Details