Product Details

SNP ID
rs142987979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45695647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTTTGTAGGGGCCACGGATTCG[G/T]CTGTGCCCTGTGCCCTGCTGCTGGA
Phenotype
MIM: 601061
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
QPCTL PubMed Links

Gene Details

Gene
QPCTL
Gene Name
glutaminyl-peptide cyclotransferase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163377.1 862 Intron NP_001156849.1
NM_017659.3 862 Missense Mutation GCT,TCT A188S NP_060129.2
XM_011527048.2 862 Missense Mutation GCT,TCT A188S XP_011525350.1
XM_017026900.1 862 Missense Mutation GCT,TCT A188S XP_016882389.1
Gene
SNRPD2
Gene Name
small nuclear ribonucleoprotein D2 polypeptide
There are no transcripts associated with this gene.

View Full Product Details