Product Details

SNP ID
rs143046006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40192347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCAGCTAGAGGAGATCATCGGT[C/G]TGGGGGGCTTTGGCAAGGTCTATCG
Phenotype
MIM: 600137
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAP3K10 PubMed Links

Gene Details

Gene
MAP3K10
Gene Name
mitogen-activated protein kinase kinase kinase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002446.3 604 Missense Mutation CTG,GTG L106V NP_002437.2
XM_006723222.2 604 Intron XP_006723285.1
XM_006723223.3 604 Intron XP_006723286.1
XM_011526981.2 604 Missense Mutation CTG,GTG L106V XP_011525283.1
XM_011526982.2 604 Missense Mutation CTG,GTG L106V XP_011525284.1

View Full Product Details