Product Details
- SNP ID
-
rs143121868
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:3586546 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAAGTGGACATGCAGAAGCTCCTG[C/G]GGGGTCAGATAGGCCTGGAGGACTT
- Phenotype
-
MIM: 608792
MIM: 605535
MIM: 188070
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GIPC3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8113232] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GIPC3
- Gene Name
- GIPC PDZ domain containing family member 3
- Gene
- HMG20B
- Gene Name
- high mobility group 20B
There are no transcripts associated with this gene.
- Gene
- TBXA2R
- Gene Name
- thromboxane A2 receptor
There are no transcripts associated with this gene.
View Full Product Details