Product Details

SNP ID
rs143195753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36514838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACATTCCTTACATGCATAGGGT[G/T]TGGTTCCTGTATGATTTTTCTGGTG
Phenotype
MIM: 613749
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF260 PubMed Links

Gene Details

Gene
ZNF260
Gene Name
zinc finger protein 260
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012756.2 1317 Missense Mutation AAA,ACA K134T NP_001012774.1
NM_001166036.1 1317 Missense Mutation AAA,ACA K134T NP_001159508.1
NM_001166037.1 1317 Missense Mutation AAA,ACA K134T NP_001159509.1
NM_001166038.1 1317 Missense Mutation AAA,ACA K134T NP_001159510.1
XM_011526897.2 1317 Missense Mutation AAA,ACA K134T XP_011525199.1
XM_017026741.1 1317 Missense Mutation AAA,ACA K134T XP_016882230.1

View Full Product Details