Product Details

SNP ID
rs143321857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35250494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGATGCCTTCTCCCCGGCCAGC[A/G]TCGACAACCAGCTCAATGCCCAGCT
Phenotype
MIM: 616582
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LSR PubMed Links

Gene Details

Gene
LSR
Gene Name
lipolysis stimulated lipoprotein receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260489.1 656 Missense Mutation ATC,GTC I145V NP_001247418.1
NM_001260490.1 656 Missense Mutation ATC,GTC I145V NP_001247419.1
NM_015925.6 656 Missense Mutation ATC,GTC I145V NP_057009.3
NM_205834.3 656 Missense Mutation ATC,GTC I145V NP_991403.1
NM_205835.3 656 Missense Mutation ATC,GTC I145V NP_991404.1
XM_005258980.1 656 Missense Mutation ATC,GTC I145V XP_005259037.1
XM_005258982.1 656 Missense Mutation ATC,GTC I145V XP_005259039.1
XM_011527026.1 656 Missense Mutation ATC,GTC I145V XP_011525328.1

View Full Product Details