Product Details

SNP ID
rs143326716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49290697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATTAGCAGCACAACTGGACACA[A/G]ATGGGGCCACAGCCAACCAAAGATG
Phenotype
MIM: 607972
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC6A16 PubMed Links

Gene Details

Gene
SLC6A16
Gene Name
solute carrier family 6 member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014037.2 2099 Silent Mutation CTG,TTG L617L NP_054756.2
XM_005258820.2 2099 Silent Mutation CTG,TTG L617L XP_005258877.1
XM_006723168.3 2099 Silent Mutation CTG,TTG L617L XP_006723231.1
XM_011526859.2 2099 Silent Mutation CTG,TTG L662L XP_011525161.2
XM_011526860.2 2099 Silent Mutation CTG,TTG L639L XP_011525162.1
XM_011526861.2 2099 Silent Mutation CTG,TTG L470L XP_011525163.1
XM_017026712.1 2099 Silent Mutation CTG,TTG L699L XP_016882201.1

View Full Product Details