Product Details
- SNP ID
-
rs143507031
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:7897658 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGGGATTACAAGCCTGAGCCACC[A/G]CACCCAGCCTCTCCCTGTATCCTCA
- Phenotype
-
MIM: 612891
MIM: 603014
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LRRC8E
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139881422] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LRRC8E
- Gene Name
- leucine rich repeat containing 8 family member E
- Gene
- MAP2K7
- Gene Name
- mitogen-activated protein kinase kinase 7
There are no transcripts associated with this gene.
View Full Product Details