Product Details

SNP ID
rs144165618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57720576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCACTACACACATAAGGCCTTT[C/T]CCCAGTGTGGACTTTCCAGTGCCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF671 PubMed Links

Gene Details

Gene
ZNF671
Gene Name
zinc finger protein 671
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321375.1 2102 Missense Mutation AAA,GAA K406E NP_001308304.1
NM_001321376.1 2102 Missense Mutation AAA,GAA K427E NP_001308305.1
NM_024833.2 2102 Missense Mutation AAA,GAA K504E NP_079109.2
XM_017027314.1 2102 Missense Mutation AAA,GAA K427E XP_016882803.1

View Full Product Details