Product Details

SNP ID
rs144169216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41761498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAGAACCCAGCGAGTGCCAACC[A/G]CAGTGACCCAGTCACCCTGAATGTC
Phenotype
MIM: 163980
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEACAM6 PubMed Links

Gene Details

Gene
CEACAM6
Gene Name
carcinoembryonic antigen related cell adhesion molecule 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002483.6 Intron NP_002474.4
XM_011526990.1 Intron XP_011525292.1

View Full Product Details