Product Details

SNP ID
rs144170900
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57547093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGACAGAGTGCTGAATGAGGTAC[A/G]TGCTCCGGTGAAAGGCTTTCCCACA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZNF549 PubMed Links

Gene Details

Gene
ZNF549
Gene Name
zinc finger protein 549
There are no transcripts associated with this gene.

Gene
ZNF550
Gene Name
zinc finger protein 550
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277090.1 1329 Missense Mutation ACG,ATG T384M NP_001264019.1
NM_001277091.1 1329 Missense Mutation ACG,ATG T384M NP_001264020.1
NM_001277092.1 1329 Missense Mutation ACG,ATG T384M NP_001264021.1
NM_001277093.1 1329 Intron NP_001264022.1
XM_011526567.2 1329 Missense Mutation ACG,ATG T384M XP_011524869.1
XM_011526568.2 1329 Missense Mutation ACG,ATG T375M XP_011524870.1
XM_017026401.1 1329 Missense Mutation ACG,ATG T384M XP_016881890.1

View Full Product Details