Product Details

SNP ID
rs144233412
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15889671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGGGCACTGAGCTCCAAGATGG[A/C]GGCAATATATTCACTGGGTTTCCTG
Phenotype
MIM: 604426
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CYP4F2 PubMed Links
Additional Information
For this assay, SNP(s) [rs3093156] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CYP4F2
Gene Name
cytochrome P450 family 4 subfamily F member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082.4 Intron NP_001073.3

View Full Product Details