Product Details

SNP ID
rs144459600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45872350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGTATCGGCGGCCCCTGGCACA[C/T]GCCAAGCCACCGTATTCCTATATCT
Phenotype
MIM: 602295 MIM: 602388
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXA3 PubMed Links

Gene Details

Gene
FOXA3
Gene Name
forkhead box A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004497.2 542 Silent Mutation CAC,CAT H115H NP_004488.2
Gene
SYMPK
Gene Name
symplekin
There are no transcripts associated with this gene.

View Full Product Details