Product Details

SNP ID
rs144549992
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57135177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTTACATTCATAGGGCTTTTCC[C/T]CAGTATGGATTTTTTTATGTTGAAT
Phenotype
MIM: 609546
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
USP29 PubMed Links

Gene Details

Gene
USP29
Gene Name
ubiquitin specific peptidase 29
There are no transcripts associated with this gene.

Gene
ZIM3
Gene Name
zinc finger imprinted 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052882.1 1546 Missense Mutation GAG,GGG E387G NP_443114.1

View Full Product Details