Product Details

SNP ID
rs144667068
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36514088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACTACACTGATAAGGTTTTTCA[C/G]CAGTATGGATTCTCATGTGCAGAGC
Phenotype
MIM: 613749
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ZNF260 PubMed Links

Gene Details

Gene
ZNF260
Gene Name
zinc finger protein 260
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012756.2 2067 Missense Mutation GCT,GGT A384G NP_001012774.1
NM_001166036.1 2067 Missense Mutation GCT,GGT A384G NP_001159508.1
NM_001166037.1 2067 Missense Mutation GCT,GGT A384G NP_001159509.1
NM_001166038.1 2067 Missense Mutation GCT,GGT A384G NP_001159510.1
XM_011526897.2 2067 Missense Mutation GCT,GGT A384G XP_011525199.1
XM_017026741.1 2067 Missense Mutation GCT,GGT A384G XP_016882230.1

View Full Product Details