Product Details

SNP ID
rs144672035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43204248 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGGATTCCAGAAGTTTAAAAGT[A/G]ATGCTAGGAGGTACAGAGAGCATCA
Phenotype
MIM: 176393
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC284344 PubMed Links

Gene Details

Gene
LOC284344
Gene Name
uncharacterized LOC284344
There are no transcripts associated with this gene.

Gene
PSG4
Gene Name
pregnancy specific beta-1-glycoprotein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276495.1 306 Missense Mutation TCA,TTA S23L NP_001263424.1
NM_001316339.1 306 Intron NP_001303268.1
NM_002780.4 306 Missense Mutation TCA,TTA S23L NP_002771.2
NM_213633.2 306 Missense Mutation TCA,TTA S23L NP_998798.1
XM_017026995.1 306 Intron XP_016882484.1
XM_017026996.1 306 Missense Mutation TCA,TTA S39L XP_016882485.1
XM_017026997.1 306 Missense Mutation TCA,TTA S23L XP_016882486.1
XM_017026998.1 306 Intron XP_016882487.1
XM_017026999.1 306 Intron XP_016882488.1
XM_017027000.1 306 Missense Mutation TCA,TTA S39L XP_016882489.1
XM_017027001.1 306 Intron XP_016882490.1
XM_017027002.1 306 Intron XP_016882491.1

View Full Product Details