Product Details

SNP ID
rs144672614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49890243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGAAAATCATGCGCGACGGGCG[A/G]TCGGTGTTTGAGTGGCCGCCTTCAA
Phenotype
MIM: 609742 MIM: 616659
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL4I1 PubMed Links

Gene Details

Gene
IL4I1
Gene Name
interleukin 4 induced 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258017.1 1760 Silent Mutation GAC,GAT D399D NP_001244946.1
NM_001258018.1 1760 Silent Mutation GAC,GAT D399D NP_001244947.1
NM_152899.1 1760 Silent Mutation GAC,GAT D377D NP_690863.1
NM_172374.2 1760 Silent Mutation GAC,GAT D399D NP_758962.1
Gene
MIR4750
Gene Name
microRNA 4750
There are no transcripts associated with this gene.

Gene
TBC1D17
Gene Name
TBC1 domain family member 17
There are no transcripts associated with this gene.

View Full Product Details