Product Details

SNP ID
rs144862480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44077357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTGTATGTTATTTACTGGTCTT[C/T]TCTTATTCCTTTATGTATTAAGGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF223 PubMed Links

Gene Details

Gene
ZNF223
Gene Name
zinc finger protein 223
There are no transcripts associated with this gene.

Gene
ZNF284
Gene Name
zinc finger protein 284
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011526907.2 Intron XP_011525209.1
XM_011526908.2 Intron XP_011525210.1

View Full Product Details