Product Details

SNP ID
rs144892283
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35225027 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTCCACTGAGCCTCCAGCGTC[C/T]CCAGACTGGCGGCGGGTTTGAACTG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAM187B PubMed Links

Gene Details

Gene
FAM187B
Gene Name
family with sequence similarity 187 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152481.1 953 Missense Mutation GAG,GGG E303G NP_689694.1

View Full Product Details