Product Details

SNP ID
rs144964911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50874862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCACCCCCAGTGGGTGCTCACAG[C/T]TGCCCATTGCCTAAAGAAGTAAGTA
Phenotype
MIM: 147960
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLK2 PubMed Links

Gene Details

Gene
KLK2
Gene Name
kallikrein related peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002231.2 229 Missense Mutation GCT,GTT A63V NP_001002231.1
NM_001256080.1 229 Intron NP_001243009.1
NM_005551.4 229 Missense Mutation GCT,GTT A63V NP_005542.1
Gene
KLKP1
Gene Name
kallikrein pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details