Product Details

SNP ID
rs144968769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46466687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTTGGCACACACCCCAGGAGAAC[A/G]TCGATGCACACAGCTGTGTAGCTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PNMAL1 PubMed Links

Gene Details

Gene
PNMAL1
Gene Name
paraneoplastic Ma antigen family like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103149.1 3585 UTR 3 NP_001096619.1
NM_018215.3 3585 UTR 3 NP_060685.2
XM_011527067.2 3585 UTR 3 XP_011525369.1

View Full Product Details