Product Details

SNP ID
rs144981297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14826495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGACCATGGCACCAAATCCATCA[C/G]TCCAATAAATAAACTTATATATGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR7A5 PubMed Links

Gene Details

Gene
OR7A5
Gene Name
olfactory receptor family 7 subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017506.1 1884 UTR 3 NP_059976.1
XM_011527912.2 1884 UTR 3 XP_011526214.1
XM_011527913.2 1884 UTR 3 XP_011526215.1
XM_017026583.1 1884 UTR 3 XP_016882072.1
Gene
OR7C1
Gene Name
olfactory receptor family 7 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198944.1 1884 Intron NP_945182.1
XM_005259853.3 1884 Intron XP_005259910.1
XM_011527915.2 1884 Intron XP_011526217.1

View Full Product Details