Product Details

SNP ID
rs145259987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17050014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGTTGTCGTCCTCAGACAGGGG[A/C]GTGTTCTTGGGTGCTCCCCCAGATT
Phenotype
MIM: 613434
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HAUS8 PubMed Links

Gene Details

Gene
HAUS8
Gene Name
HAUS augmin like complex subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011699.1 1203 Silent Mutation ACG,ACT T363T NP_001011699.1
NM_033417.1 1203 Silent Mutation ACG,ACT T364T NP_219485.1
XM_005260154.4 1203 Silent Mutation ACG,ACT T219T XP_005260211.1

View Full Product Details