Product Details

SNP ID
rs145266047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46411535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGGGCCTGTGCCCTCTGATTAT[C/T]TGCACCCTGGACAGCTGGGGCCCTT
Phenotype
MIM: 614145
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC8 PubMed Links

Gene Details

Gene
CCDC8
Gene Name
coiled-coil domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032040.4 2128 Missense Mutation NP_114429.2

View Full Product Details