Product Details

SNP ID
rs145336850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:35891343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGCTGATAACAGGGCAAAACCA[C/T]CTAGTTTGCTCGTCCAAAAAATGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NFKBID PubMed Links

Gene Details

Gene
NFKBID
Gene Name
NFKB inhibitor delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321831.1 Intron NP_001308760.1
NM_139239.2 Intron NP_640332.1
XM_005259340.3 Intron XP_005259397.1
XM_011527419.2 Intron XP_011525721.1
XM_011527420.2 Intron XP_011525722.1
XM_011527421.2 Intron XP_011525723.1
XM_011527424.2 Intron XP_011525726.1
XM_017027389.1 Intron XP_016882878.1
XM_017027390.1 Intron XP_016882879.1
XM_017027391.1 Intron XP_016882880.1
XM_017027392.1 Intron XP_016882881.1
XM_017027393.1 Intron XP_016882882.1
XM_017027394.1 Intron XP_016882883.1
XM_017027395.1 Intron XP_016882884.1
XM_017027396.1 Intron XP_016882885.1
XM_017027397.1 Intron XP_016882886.1

View Full Product Details