Product Details

SNP ID
rs145362201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39200413 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGGCTGCTGGCGGCCGACCGC[C/T]GCACGGTCATTGCTCAGCACCACGT
Phenotype
MIM: 615901
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCCRP1 PubMed Links

Gene Details

Gene
NCCRP1
Gene Name
non-specific cytotoxic cell receptor protein 1 homolog (zebrafish)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001414.1 635 Missense Mutation CGC,TGC R206C NP_001001414.1
XM_011526906.2 635 Intron XP_011525208.1
Gene
SYCN
Gene Name
syncollin
There are no transcripts associated with this gene.

View Full Product Details