Product Details

SNP ID
rs145419216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11576517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACATAGTGTCACTGTGTCCAGC[A/G]TAAAAATGGCCACAGACACATTGGT
Phenotype
MIM: 171640 MIM: 612248
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACP5 PubMed Links

Gene Details

Gene
ACP5
Gene Name
acid phosphatase 5, tartrate resistant
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111034.2 864 Missense Mutation ACG,ATG T154M NP_001104504.1
NM_001111035.2 864 Missense Mutation ACG,ATG T154M NP_001104505.1
NM_001111036.2 864 Missense Mutation ACG,ATG T154M NP_001104506.1
NM_001322023.1 864 Missense Mutation ACG,ATG T154M NP_001308952.1
NM_001611.4 864 Missense Mutation ACG,ATG T154M NP_001602.1
XM_005259938.1 864 Missense Mutation ACG,ATG T154M XP_005259995.1
XM_011528069.2 864 Missense Mutation ACG,ATG T154M XP_011526371.1
Gene
ZNF627
Gene Name
zinc finger protein 627
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290083.1 864 Intron NP_001277012.1
NM_001290084.1 864 Intron NP_001277013.1
NM_001290085.1 864 Intron NP_001277014.1
NM_145295.3 864 Intron NP_660338.1
XM_011527780.2 864 Intron XP_011526082.1

View Full Product Details