Product Details

SNP ID
rs145464132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:677832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGCAGGGCCAGGAGGCCACCTG[C/T]AGCCTGGTGCTCCAGACTGATGTCA
Phenotype
MIM: 605343
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FSTL3 PubMed Links

Gene Details

Gene
FSTL3
Gene Name
follistatin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005860.2 179 Silent Mutation TGC,TGT C48C NP_005851.1
Gene
PRSS57
Gene Name
protease, serine 57
There are no transcripts associated with this gene.

View Full Product Details