Product Details
- SNP ID
-
rs145548575
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:40423297 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTAGGCACAGGTGGCAGGGCAGCCG[C/T]GGGTGCCATGGACGCCTGGATGCGC
- Phenotype
-
MIM: 605725
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRX
PubMed Links
Gene Details
- Gene
- PRX
- Gene Name
- periaxin
There are no transcripts associated with this gene.
- Gene
- SERTAD1
- Gene Name
- SERTA domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013376.3 |
409 |
Missense Mutation |
ACG,GCG |
T84A |
NP_037508.2 |
View Full Product Details