Product Details

SNP ID
rs145804391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17250404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCGGTGGGCTCCTCGGAAGGCCA[A/G]GACCAGGGCGCTGGAAGGGGTGGGT
Phenotype
MIM: 132880 MIM: 611810
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NR2F6 PubMed Links

Gene Details

Gene
NR2F6
Gene Name
nuclear receptor subfamily 2 group F member 6
There are no transcripts associated with this gene.

Gene
USHBP1
Gene Name
USH1 protein network component harmonin binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297703.1 1958 Silent Mutation CTG,TTG L581L NP_001284632.1
NM_001321417.1 1958 Silent Mutation CTG,TTG L645L NP_001308346.1
NM_031941.3 1958 Silent Mutation CTG,TTG L645L NP_114147.2
XM_011528351.2 1958 Intron XP_011526653.1

View Full Product Details