Product Details

SNP ID
rs145925971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14950210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGGCCCCTCACATAGCACTCTC[A/G]TTGCCTCCCCGTCCCCGGGATGCCC
Phenotype
MIM: 600637
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR7C2 PubMed Links

Gene Details

Gene
OR7C2
Gene Name
olfactory receptor family 7 subfamily C member 2
There are no transcripts associated with this gene.

Gene
SLC1A6
Gene Name
solute carrier family 1 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272087.1 1871 Intron NP_001259016.1
NM_001272088.1 1871 Intron NP_001259017.1
NM_005071.2 1871 Silent Mutation AAC,AAT N560N NP_005062.1
XM_006722842.1 1871 Silent Mutation AAC,AAT N560N XP_006722905.1
XM_006722844.1 1871 Silent Mutation AAC,AAT N560N XP_006722907.1
XM_017027152.1 1871 Silent Mutation AAC,AAT N560N XP_016882641.1

View Full Product Details